Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18154

Sigma-Aldrich

Trichloromethyl chloroformate

for synthesis

Synonym(s):

Trichloromethyl chloroformate, Diphosgene, Chloroformic acid trichloromethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
197.83
MDL number:
UNSPSC Code:
12352108
EC Index Number:
207-965-9
NACRES:
NA.22
grade:
for synthesis
bp:
128 °C/1.013 hPa
vapor pressure:
13 hPa ( 20 °C)

grade

for synthesis

Quality Level

vapor pressure

13 hPa ( 20 °C)

form

liquid

dilution

(for synthesis)

bp

128 °C/1.013 hPa

mp

-57 °C

density

1.63 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

ClC(Cl)(Cl)OC(=O)Cl

InChI

1S/C2Cl4O2/c3-1(7)8-2(4,5)6

InChI key

HCUYBXPSSCRKRF-UHFFFAOYSA-N

General description

for synthesis

Application

Trichloromethyl chloroformate for synthesis. CAS 503-38-8, EC Number 207-965-9, chemical formula ClCOOCCl₃.

Analysis Note

Assay (argentometric)≥ 98.0 %Density (d 20 °C/ 4 °C)1.635 - 1.642Identity (IR)passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service