Skip to Content
MilliporeSigma

Skip To

8.17007

Polyethylene glycol

6000, EMPROVE® ESSENTIAL, Ph. Eur., NF

Synonym(s):

Poly(ethylene glycol), Polyglycol, Polyethylene oxide, Polyoxy ethylene, PEG 6000, PEG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
UNSPSC Code:
13111021
EC Index Number:
500-038-2
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

NF, Ph. Eur.

vapor pressure

<0.01 hPa ( 20 °C)

product line

EMPROVE® ESSENTIAL

form

solid

autoignition temp.

420 °C

mol wt

average Mn 6000

technique(s)

API processing | nano-milling: suitable

pH

5-7 (20 °C, 100 g/L in H2O)

mp

58-63 °C

transition temp

flash point 270 °C

solubility

550 g/L

density

1.2 g/cm3 at 20 °C

application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation
solubility enhancement

storage temp.

15-25°C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

General description

When working on a pharmaceutical formulation, there are specific considerations to make and challenging hurdles to take before you can successfully launch your final drug product - formulation stability, release kinetics and bioavailability limitations just to name a few. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Polyethylene glycol (PEG) is a multipurpose polymer which can be applied in solid, semi-solid and liquid formulations. Its broad functional spectrum includes e.g. improvement of API solubility, acting as a lubricant for tablet coating, function as a carrier, and many more. Our comprehensive portfolio of PEGs comprises diverse polymer sizes of different molecular weights to meet your specific application and needs.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.170198.170188.17003
application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation
solubility enhancement

application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation
solubility enhancement

application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation
solubility enhancement

application(s)

liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation
solubility enhancement

technique(s)

API processing | nano-milling: suitable

technique(s)

API processing | nano-milling: suitable

technique(s)

API processing | nano-milling: suitable

technique(s)

-

agency

NF, Ph. Eur.

agency

NF, Ph. Eur.

agency

NF, Ph. Eur.

agency

JP, NF, Ph. Eur.

form

solid

form

scales

form

scales

form

liquid

product line

EMPROVE® ESSENTIAL

product line

EMPROVE® ESSENTIAL

product line

EMPROVE® ESSENTIAL

product line

EMPROVE® EXPERT

solubility

550 g/L

solubility

-

solubility

500 g/L

solubility

-


Still not finding the right product?

Explore all of our products under Polyethylene glycol


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

281.5 °F - closed cup

flash_point_c

138.6 °C - closed cup



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters