Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.16144

Sigma-Aldrich

Oxalic acid

anhydrous for synthesis

Synonym(s):

Oxalic acid, Ethanedioic acid dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H2O4
CAS Number:
Molecular Weight:
90.03
MDL number:
UNSPSC Code:
12352106
EC Index Number:
205-634-3
NACRES:
NA.22
pH:
1 (20 °C, 100 g/L in H2O)

vapor pressure

<1 Pa ( 25 °C)

Quality Level

Assay

≥99% (manganometric)

form

solid

potency

375 mg/kg LD50, oral (Rat)

pH

1 (20 °C, 100 g/L in H2O)

mp

189 °C (decomposition)

solubility

108 g/L

density

1.9 g/cm3 at 20 °C

bulk density

750 kg/m3

storage temp.

2-30°C

SMILES string

[O+H2]C(=O)C(=O)[O-]

InChI

1S/C2H2O4/c3-1(4)2(5)6/h(H,3,4)(H,5,6)

InChI key

MUBZPKHOEPUJKR-UHFFFAOYSA-N

Application

  • Glycerol conversion into added-value products on Ni-Cu based catalysts: Discusses the role of oxalic acid among various catalysts in glycerol conversion to value-added products (JLC Fajín, MNDS Cordeiro, 2024).
  • Synthesis, Crystal Structure, Hirshfeld Surface and Computational Analysis of Bis-Oxalate Ethanol Solvate: Explores the crystal structure and properties of a new oxalate compound (P Solo, M Arockia doss, 2024).
  • Synergistic Enhancement of Barrier Performance in Silanized Modified Hemicellulose/PVA Composite Films through Oxalic Acid Crosslinking: Discusses the enhancement of material properties through oxalic acid treatment (J Wang et al., 2024).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service