Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.16020

Sigma-Aldrich

(S)-(+)-Serine

for synthesis

Synonym(s):

(S)-(+)-Serine, 3-Hydroxy-L-alanine, Ser

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7NO3
CAS Number:
Molecular Weight:
105.09
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-274-3
NACRES:
NA.22

Quality Level

form

solid

pH

5-6 (20 °C, 100 g/L in H2O)

mp

215-225 °C (decomposes)

solubility

360 g/L

bulk density

570 kg/m3

storage temp.

2-30°C

SMILES string

[N+H3]C(CO)C(=O)[O-]

InChI

1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)

InChI key

MTCFGRXMJLQNBG-UHFFFAOYSA-N

General description

(S)-(+)-Serine is a non-essential amino acid that serves as a building block in peptide synthesis and as a precursor for various biomolecules, including neurotransmitters and phospholipids.

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Spec. rotation [α²0/D (c=10 in HCl 2 mol/l): +14.0 - +15.5 °
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service