Skip to Content
MilliporeSigma

8.14971

Di-(N,N′-succinimidyl) carbonate

for synthesis

Synonym(s):

Di-(N,N′-succinimidyl) carbonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H8N2O7
CAS Number:
Molecular Weight:
256.17
UNSPSC Code:
12352108
EC Index Number:
277-730-3
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (HPLC)

form

solid

reaction suitability

reaction type: Carbonylations

mp

188-191 °C

application(s)

peptide synthesis

storage temp.

2-30°C

SMILES string

N2(C(=O)CCC2=O)OC(=O)ON1C(=O)CCC1=O

InChI

1S/C9H8N2O7/c12-5-1-2-6(13)10(5)17-9(16)18-11-7(14)3-4-8(11)15/h1-4H2

InChI key

PFYXSUNOLOJMDX-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test


pictograms

Exclamation markHealth hazard

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT RE 2 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library