Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14926

Sigma-Aldrich

Methyl heptanoate

for synthesis

Synonym(s):

Methyl heptanoate, Methyl enanthate, Heptanoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O2
CAS Number:
Molecular Weight:
144.21
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-428-8
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

bp

173 °C/1013 hPa

mp

-56 °C

transition temp

flash point 57 °C

density

0.880 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)C(=O)CCCCCC

InChI

1S/C8H16O2/c1-3-4-5-6-7-8(9)10-2/h3-7H2,1-2H3

InChI key

XNCNNDVCAUWAIT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.878 - 0.882
Identity (IR): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service