Skip to Content
MilliporeSigma

Skip To

8.14737

DL-Malic acid

for synthesis

Synonym(s):

DL-Malic acid, DL-Hydroxysuccinic acid, Hydroxybutanedioic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C4H6O5
CAS Number:
Molecular Weight:
134.09
UNSPSC Code:
12352106
EC Index Number:
230-022-8
NACRES:
NA.22
MDL number:
Assay:
≥99.0% (acidimetric)
Form:
solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99.0% (acidimetric)

form

solid

autoignition temp.

349 °C

potency

>3200 mg/kg LD50, oral (Rat)

pH

2.3 (20 °C, 10 g/L in H2O)

mp

128-132 °C

transition temp

flash point 203 °C

solubility

558 g/L

density

1.60 g/cm3 at 20 °C

bulk density

800 kg/m3

storage temp.

2-30°C

SMILES string

[O-]C(=O)C(O)CC(=O)[O-].[H+].[H+]

InChI

1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)

InChI key

BJEPYKJPYRNKOW-UHFFFAOYSA-N

Application

  • Study on the inhibitory effect of DL‐malic acid on the phase structure and combustion characteristics of coal: This study examines the potential of DL-malic acid as an antioxidant inhibitor in the prevention of coal fires (Sun et al., 2024).

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Melting range (lower value): ≥ 128 °C
Melting range (upper value): ≤ 132 °C
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
100382100383240176
description

for synthesis

description

for biochemistry

description

EMPROVE® ESSENTIAL, Ph. Eur., JPE, NF, FCC, E 296

description

ReagentPlus®, ≥99%

assay

≥99.0% (acidimetric)

assay

≥99.5% (acidimetric)

assay

-

assay

≥99%

solubility

558 g/L

solubility

-

solubility

558 g/L

solubility

acetone: soluble 17.75g/100g at 20 °C(lit.), diethyl ether: soluble 0.84g/100g at 20 °C(lit.), dioxane: soluble 22.7 g/100g at 20 °C(lit.), ethanol: soluble 45.53g/100g at 20 °C(lit.), methanol: soluble 82.7 g/100g at 20 °C(lit.), water: soluble 55.8g/100g at 20 °C(lit.), benzene: insoluble(lit.)

Quality Level

200

Quality Level

100

Quality Level

500

Quality Level

200

density

1.60 g/cm3 at 20 °C

density

1.60 g/cm3 at 20 °C

density

1.60 g/cm3 at 20 °C

density

-

form

solid

form

solid

form

solid

form

solid

mp

128-132 °C

mp

128-132 °C

mp

128-132 °C

mp

131-133 °C (lit.)


Still not finding the right product?

Explore all of our products under DL-Malic acid


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

397.4 °F

flash_point_c

203 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters