Skip to Content
MilliporeSigma

8.14725

Toluene-4-sulfonic acid monohydrate

for synthesis

Synonym(s):

Toluene-4-sulfonic acid monohydrate, p-Toluenesulfonic acid, Tosylic acid, Tosic acid, PTSA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H8O3S · xH2O
CAS Number:
Molecular Weight:
172.20 (anhydrous basis)
UNSPSC Code:
12352106
EC Index Number:
203-180-0
NACRES:
NA.22
MDL number:
Grade:
synthesis grade
Form:
crystals
Solubility:
750 g/L
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Segment

form

crystals

autoignition temp.

600 °C

potency

2570 mg/kg LD50, oral (Rat)

pH

1 (20 °C, 650 g/L in H2O, anhydrous substance)

bp

140 °C/27 hPa

mp

56 °C (Elimination of water of crystallisation)

transition temp

flash point 180 °C

solubility

750 g/L

bulk density

510 kg/m3

storage temp.

2-30°C

SMILES string

[S](=O)(=O)([O-])c1ccc(cc1)C.[O+H3]

InChI

1S/C7H8O3S.H2O/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2

InChI key

KJIFKLIQANRMOU-UHFFFAOYSA-N

Application

  • Synthesis and characterization of toluene sulfonic acid (tsa) doped polypyrrole nanoparticles: Effect of dopant concentrations: This study investigates the effect of various concentrations of para-toluene sulfonic acid on the properties of polypyrrole nanoparticles, providing insights into the optimization of conductive polymers for applications in electronic devices and sensors (Alva et al., 2016).

Analysis Note

Assay (acidimetric, calc. on anhydrous substance): ≥ 98.0 %
Water (K. F.): 8.0 - 12.0 %
Identity (IR): passes test


Still not finding the right product?


target_organs

Respiratory system

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

8A - Combustible corrosive hazardous materials



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library