Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14440

Sigma-Aldrich

3-Phenylpropyl acetate

for synthesis

Synonym(s):

3-Phenylpropyl acetate, Hydroxycinnamyl acetate, ß-Phenylpropyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14O2
CAS Number:
Molecular Weight:
178.23
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-569-8
NACRES:
NA.22

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

autoignition temp.

445 °C

potency

4700 mg/kg LD50, oral (Rat)

bp

245 °C

transition temp

flash point 130 °C

solubility

0.69 g/L

density

1.01 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C11H14O2/c1-10(12)13-9-5-8-11-6-3-2-4-7-11/h2-4,6-7H,5,8-9H2,1H3

InChI key

JRJGKUTZNBZHNK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.014 - 1.016
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service