Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
(CH3)2C(OCH3)CH2CH2OH
CAS Number:
Molecular Weight:
118.17
NACRES:
NA.22
UNSPSC Code:
12191601
Beilstein/REAXYS Number:
1732882
MDL number:
vapor pressure
0.66 hPa ( 25 °C)
Quality Segment
greener alternative product characteristics
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
refractive index
n
bp
173-175 °C (lit.)
mp
<-50 °C
density
0.926 g/mL at 20 °C (lit.)
greener alternative category
SMILES string
COC(C)(C)CCO
InChI
1S/C6H14O2/c1-6(2,8-3)4-5-7/h7H,4-5H2,1-3H3
InChI key
MFKRHJVUCZRDTF-UHFFFAOYSA-N
General description
3-Methoxy-3-methyl-1-butanol, considered as an eco-friendly solvent, is widely used in paints, inks, fragrances, air fresheners, household, and industrial cleaner formulations due to its non-toxic and non-volatile characteristics. It can also be used as a raw material to produce industrial detergents.
Features and Benefits
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is considered as a multifunctional Chemical for a cleaner and Greener Environment and thus aligns with "Safer Solvents and Auxiliaries". Click here for more information.
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class
10 - Combustible liquids
WGK
WGK 1
Flash Point (°F)
159.8 °F - closed cup
Flash Point (°C)
71 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
