Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14269

Sigma-Aldrich

Methylphenyl glyoxylate

for synthesis

Synonym(s):

Methylphenyl glyoxylate, Phenylglyoxylic acid methyl ester, Benzoyl formic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8O3
CAS Number:
Molecular Weight:
164.16
MDL number:
UNSPSC Code:
12352108
EC Index Number:
239-263-3
NACRES:
NA.22

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

>500 °C

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rat)

mp

17 °C (ECHA)

transition temp

flash point >113 °C

density

1.16 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)C(=O)C(=O)c1ccccc1

InChI

1S/C9H8O3/c1-12-9(11)8(10)7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YLHXLHGIAMFFBU-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.159 - 1.163
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service