Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.08384

Sigma-Aldrich

DL-Tropic acid

for synthesis

Synonym(s):

DL-Tropic acid, 3-Hydroxy-2-phenylpropionic acid, α-Phenylhydracrylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(C6H5)COOH
CAS Number:
Molecular Weight:
166.17
MDL number:
UNSPSC Code:
12352106
EC Index Number:
208-465-3
NACRES:
NA.22

Quality Level

Assay

≥97.0% (acidimetric)

form

powder

mp

115-118 °C

solubility

20 g/L

storage temp.

2-30°C

SMILES string

OCC(c1ccccc1)C(=O)O

InChI

1S/C9H10O3/c10-6-8(9(11)12)7-4-2-1-3-5-7/h1-5,8,10H,6H2,(H,11,12)

InChI key

JACRWUWPXAESPB-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (acidimetric): ≥ 97.0 %
Melting range (lower value): ≥ 115 °C
Melting range (upper value): ≤ 118 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service