Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.08278

Sigma-Aldrich

Tetra-n-butyltin

for synthesis

Synonym(s):

Tetra-n-butyltin, Tetrabutylstannane, Tin tetrabutyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4Sn
CAS Number:
Molecular Weight:
347.17
MDL number:
UNSPSC Code:
12352103
EC Index Number:
215-960-8
NACRES:
NA.22

vapor pressure

0.0014 hPa ( 25 °C)

Quality Level

Assay

≥95.0% (GC)

form

liquid

potency

1268 mg/kg LD50, oral (Rat)

bp

140 °C/10 hPa

mp

-97 °C

transition temp

flash point >100 °C

solubility

0.008 g/L

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Sn](CCCC)(CCCC)(CCCC)CCCC

InChI

1S/4C4H9.Sn/c4*1-3-4-2;/h4*1,3-4H2,2H3;

InChI key

AFCAKJKUYFLYFK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.052 - 1.057
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

231.8 - 239.0 °F - closed cup

Flash Point(C)

111 - 115 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service