Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.08230

Sigma-Aldrich

4-Toluenesulfonamide

for synthesis

Synonym(s):

4-Toluenesulfonamide, PTSA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(CH3)C6H4SO2NH2
CAS Number:
Molecular Weight:
171.22
MDL number:
UNSPSC Code:
12352111
EC Index Number:
200-741-1
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (HPLC)

form

crystals

bp

220-222 °C/13 hPa

mp

136-138 °C

transition temp

flash point 202 °C

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(N)c1ccc(cc1)C

InChI

1S/C7H9NO2S/c1-6-2-4-7(5-3-6)11(8,9)10/h2-5H,1H3,(H2,8,9,10)

InChI key

LMYRWZFENFIFIT-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 136 °C
Melting range (upper value): ≤ 140 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

395.6 °F - closed cup

Flash Point(C)

202 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service