Skip to Content
MilliporeSigma

8.07500

n-Amyl alcohol

for synthesis

Synonym(s):

n-Amyl alcohol, 1-Pentanol, Pentyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)4OH
CAS Number:
Molecular Weight:
88.15
UNSPSC Code:
12352104
EC Index Number:
200-752-1
NACRES:
NA.22
MDL number:
Assay:
≥98% (GC)
Grade:
synthesis grade
Bp:
138 °C/1013 hPa
Vapor pressure:
2.04 hPa ( 20 °C)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Segment

vapor pressure

2.04 hPa ( 20 °C)

assay

≥98% (GC)

form

liquid

autoignition temp.

300 °C (DIN 51794)

potency

3670 mg/kg LD50, oral (Rat), 2306 mg/kg LD50, skin (Rabbit)

expl. lim.

1.6 % (v/v)

dilution

(for synthesis)

pH

7 (20 °C, 22 g/L)

bp

138 °C/1013 hPa

mp

-78 °C

transition temp

flash point 47 °C

solubility

22 g/L

density

0.815 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

OCCCCC

InChI

1S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3

InChI key

AMQJEAYHLZJPGS-UHFFFAOYSA-N

Application


  • A Compact and Smooth CH₃NH₃PbI₃ Film: Investigation of Solvent Sorts and Concentrations of CH₃NH₃I towards Highly Efficient Perovskite Solar Cells.: This article investigates the role of various solvents, including n-amyl alcohol, in producing highly efficient perovskite solar cells, contributing to advancements in solar energy technology (Chen et al., 2018).


  • A novel mechanism for red emission carbon dots: hydrogen bond dominated molecular states emission.: The study identifies the use of n-amyl alcohol in synthesizing red-emission carbon dots, elucidating a new mechanism dominated by hydrogen bonding and its potential applications in bioimaging and optoelectronics (Zhang et al., 2017).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.814 - 0.815
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under n-Amyl alcohol


signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

flash_point_f

120.2 °F - closed cup

flash_point_c

49 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library