Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.07000

Sigma-Aldrich

o-Phenetidine

for synthesis

Synonym(s):

o-Phenetidine, 2-Ethoxyaniline, 2′-Aminophenetol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(C2H5O)C6H4NH2
CAS Number:
Molecular Weight:
137.18
MDL number:
UNSPSC Code:
12352116
EC Index Number:
202-356-4

vapor pressure

1 hPa ( 67 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

425 °C

bp

232-234 °C/1013 hPa

mp

-4 °C

transition temp

flash point 112 °C

density

1.05 g/cm3 at 20 °C

storage temp.

15-25°C

InChI

1S/C8H11NO/c1-2-10-8-6-4-3-5-7(8)9/h3-6H,2,9H2,1H3

InChI key

ULHFFAFDSSHFDA-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.048 - 1.050
Identity (IR): passes test

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - STOT RE 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service