Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.06914

Sigma-Aldrich

(R)-(-)-Mandelic acid

for resolution of racemates for synthesis

Synonym(s):

(R)-(-)-Mandelic acid, D-Mandelic acid, D-α-Hydroxyphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)COOH
CAS Number:
Molecular Weight:
152.15
MDL number:
UNSPSC Code:
12352106
EC Index Number:
210-276-6
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (acidimetric)

form

crystals

potency

5000 mg/kg LD50, oral (Rat)

pH

2.1 (20 °C, 60 g/L in H2O)

mp

130-132 °C

transition temp

flash point >190 °C

solubility

soluble 109.8 g/L

bulk density

670 kg/m3

storage temp.

2-30°C

SMILES string

O[C@H](c1ccccc1)C(=O)O

InChI

1S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9H,(H,10,11)/t7-/m1/s1

InChI key

IWYDHOAUDWTVEP-SSDOTTSWSA-N

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Melting range (lower value): ≥ 131 °C
Melting range (upper value): ≤ 134 °C
Spec. rotation [α²0/D (c=2 in water): -157.0 - -153.0 °
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service