Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.06862

Sigma-Aldrich

1-Naphthylacetic acid

for synthesis

Synonym(s):

1-Naphthylacetic acid, 1-Naphthaleneacetic acid, Planofix

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10O2
CAS Number:
Molecular Weight:
186.21
MDL number:
UNSPSC Code:
12352106
EC Index Number:
201-705-8
NACRES:
NA.22

Quality Level

Assay

≥96.0% (GC)

form

solid

potency

1000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

pH

3.0 (25 °C, 0.6 g/L in H2O)

mp

125-131 °C

solubility

0.42 g/L

bulk density

240 kg/m3

storage temp.

2-30°C

SMILES string

OC(=O)Cc1c2c(ccc1)cccc2

InChI

1S/C12H10O2/c13-12(14)8-10-6-3-5-9-4-1-2-7-11(9)10/h1-7H,8H2,(H,13,14)

InChI key

PRPINYUDVPFIRX-UHFFFAOYSA-N

Application

  • Syntheses, Crystal Structures, Antimicrobial Activity and Thermal Behavior of Copper (II) Complexes Derived from 1-Naphthylacetic Acid and Diamines: This study explores the structural and antimicrobial properties of copper complexes derived from 1-naphthylacetic acid (LW Xue, C Chen, GQ Zhao, 2020).
  • Simultaneous intercalation of 1-naphthylacetic acid and indole-3-butyric acid into layered double hydroxides and controlled release properties: Discusses a formulation for controlled release of 1-naphthylacetic acid in agricultural applications (S Li, Y Shen, M Xiao, D Liu, L Fan, Z Zhang, 2014).

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Melting range (lower value): ≥ 126 °C
Melting range (upper value): ≤ 134 °C
2-Naphthylacetic acid (GC, area%): ≤ 4.0 % (a/a)
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service