Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.05958

Sigma-Aldrich

Manganese(IV) oxide

(precipitated active) for synthesis

Synonym(s):

Manganese(IV) oxide, Manganese dioxide, Pyrolusite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MnO2
CAS Number:
Molecular Weight:
86.94
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-202-6
NACRES:
NA.22
grade:
for synthesis
synthesis grade
form:
powder
solubility:
insoluble <0.001 g/L

grade

for synthesis
synthesis grade

Quality Level

form

powder

potency

>3478 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: oxidant

solubility

insoluble <0.001 g/L

density

5.21 g/cm3 at 21 °C

bulk density

600‑800 kg/m3

storage temp.

2-30°C

SMILES string

[Mn](=O)=O

InChI

1S/Mn.2O

InChI key

NUJOXMJBOLGQSY-UHFFFAOYSA-N

Application

Manganese(IV) oxide (MnO2) can be used as an oxidizing agent for the oxidation of:
  • Allylic alcohols to α, β-ethylenic aldehydes, or ketones.
  • Propargylic alcohols to alkynic aldehydes or ketones.
  • Benzylic and heterocyclic alcohols to conjugated aromatic aldehydes or ketones.
  • Amines to aldehydes, imines, amides, and diazo compounds. Methyl arenes to form primary amides in the presence of urea and oxygen.
  • MnO2 can also be used to synthesize disubstituted oxazolines from aldoximes via in situ nitrile oxide formation. Non-activated cyclic and acyclic alcohols oxidized to ketone analogs using stoichiometric MnO2 and a ruthenium catalyst.

Analysis Note

Assay (iodometric, calc. on dried substance): ≥ 90.0 %
Loss on drying: ≤ 3.5 %
Oxidation Activity: passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service