Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.05650

Sigma-Aldrich

2,6-Dimethylpyridine

for synthesis

Synonym(s):

2,6-Dimethylpyridine, 2,6-Lutidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2,6-(CH3)2C5H3N
CAS Number:
Molecular Weight:
107.15
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-587-3
NACRES:
NA.22

vapor pressure

5.5 hPa ( 20 °C)

Quality Level

Assay

≥98% (GC)

form

liquid

potency

400 mg/kg LD50, oral (Rat)
>1000 mg/kg LD50, skin (Rabbit)

bp

144 °C/1013 hPa

mp

-6 °C

transition temp

flash point 33 °C

density

0.923 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

n1c(cccc1C)C

InChI

1S/C7H9N/c1-6-4-3-5-7(2)8-6/h3-5H,1-2H3

InChI key

OISVCGZHLKNMSJ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98 %
Density (d 20 °C/ 4 °C): 0.922 - 0.924
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F

Flash Point(C)

33 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service