Skip to Content
MilliporeSigma

Skip To

8.04324

1,1,1,3,3,3-Hexamethyldisilazane

for synthesis

Synonym(s):

1,1,1,3,3,3-Hexamethyldisilazane, Bis(trimethylsilyl)amine, HMDS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H19NSi2
CAS Number:
Molecular Weight:
161.39
UNSPSC Code:
12352103
EC Index Number:
213-668-5
NACRES:
NA.22
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

20 hPa ( 20 °C)

Quality Segment

assay

≥98% (GC)

form

liquid

autoignition temp.

325 °C

potency

847 mg/kg LD50, oral (Rat), 550 mg/kg LD50, skin (Rabbit)

expl. lim.

0.8-25.9 % (v/v)

kinematic viscosity

0.9 cSt(25 °C)

bp

126 °C/1013 mbar

mp

-82 °C

transition temp

flash point 15 °C

density

0.78 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Si](N[Si](C)(C)C)(C)(C)C

InChI

1S/C6H19NSi2/c1-8(2,3)7-9(4,5)6/h7H,1-6H3

InChI key

FFUAGWLWBBFQJT-UHFFFAOYSA-N

Application

  • Handling and Safety Protocols of 1,1,1,3,3,3-Hexamethyldisilazane: This article presents a comprehensive overview of the handling and safety protocols associated with 1,1,1,3,3,3-Hexamethyldisilazane, emphasizing its applications in various industrial and pharmaceutical contexts. The study provides insights on its safe usage, particularly focusing on its role as a silylation agent in analytical chemistry and as a precursor in chemical vapor deposition processes (Toxicol Ind Health, 2019).

Analysis Note

Assay (GC, area%): ≥ 98 %
Density (d 20 °C/ 4 °C): 0.774 - 0.775
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under 1,1,1,3,3,3-Hexamethyldisilazane


pictograms

FlameSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

52.5 °F - closed cup

flash_point_c

11.4 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters