Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.03605

Sigma-Aldrich

Diethyl ketone

for synthesis

Synonym(s):

Diethyl ketone, 3-Pentanone, Ethyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2CO
CAS Number:
Molecular Weight:
86.13
MDL number:
UNSPSC Code:
12352115
EC Index Number:
202-490-3
NACRES:
NA.22
grade:
for synthesis
bp:
101-103 °C/1013 mbar
vapor pressure:
37.6 hPa ( 20 °C)

grade

for synthesis

Quality Level

vapor pressure

37.6 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

425 °C

potency

2900 mg/kg LD50, oral (Rat)
16200 mg/kg LD50, skin (Rabbit)

expl. lim.

1.6-7.7 % (v/v)

dilution

(for synthesis)

pH

6.2 (20 °C, 50 g/L in H2O)

bp

101-103 °C/1013 mbar

mp

-40 °C

transition temp

flash point 7 °C

solubility

50 g/L

density

0.81 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C(CC)CC

InChI

1S/C5H10O/c1-3-5(6)4-2/h3-4H2,1-2H3

InChI key

FDPIMTJIUBPUKL-UHFFFAOYSA-N

Application


  • Resonance Stabilization Effects on Ketone Autoxidation: Isomer-Specific Cyclic Ether and Ketohydroperoxide Formation in the Low-Temperature (400-625 K) Oxidation of Diethyl Ketone.: This study examines the low-temperature oxidation of diethyl ketone, highlighting how resonance stabilization influences the formation of specific cyclic ethers and ketohydroperoxides, vital for understanding combustion mechanisms and environmental implications of ketone use in industrial applications (Scheer AM et al., 2016).

  • Low-temperature combustion chemistry of novel biofuels: resonance-stabilized QOOH in the oxidation of diethyl ketone.: This research focuses on the combustion chemistry of diethyl ketone as a biofuel, providing insights into the formation of resonance-stabilized QOOH intermediates which are crucial for developing more efficient and cleaner burning fuels (Scheer AM et al., 2014).

  • Wet effluent diffusion denuder technique and determination of volatile organic compounds in air. I. Oxo compounds (alcohols and ketones).: Explores the use of diethyl ketone in air quality monitoring, employing a novel wet effluent diffusion denuder technique for the sensitive detection of oxo compounds, vital for environmental monitoring and regulatory compliance (Pesková J et al., 2001).

  • Identification limits for volatile organic compounds in the blood by purge-and-trap GC-FTIR.: Discusses the detection limits of volatile organic compounds, including diethyl ketone, in blood using advanced purge-and-trap gas chromatography coupled with Fourier-transform infrared spectroscopy, emphasizing its importance in forensic and toxicological analyses (Ojanperä I et al., 1998).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.813 - 0.815
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

44.6 °F

Flash Point(C)

7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service