Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.03296

Sigma-Aldrich

Epichlorohydrine

for synthesis

Synonym(s):

Epichlorohydrine, 1-Chloro-2,3-epoxypropane, 2-Chloromethyl oxirane, 2,3-Epoxypropyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5ClO
CAS Number:
Molecular Weight:
92.52
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-439-8
NACRES:
NA.22

vapor pressure

16.5 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

385 °C

potency

90 mg/kg LD50, oral (Rat)
515 mg/kg LD50, skin (Rabbit)

expl. lim.

3.8-21 % (v/v)

mp

-57 °C

transition temp

flash point 28 °C

solubility

65.9 g/L

density

1.18 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C3H5ClO/c4-1-3-2-5-3/h3H,1-2H2

InChI key

BRLQWZUYTZBJKN-UHFFFAOYSA-N

Application

  • Water-insoluble β-cyclodextrin–epichlorohydrin polymers for removal of pollutants: This review article discusses the use of β-cyclodextrin–epichlorohydrin polymers in sorption processes to remove pollutants from aqueous solutions (N Morin-Crini, P Winterton, S Fourmentin, 2018, Progress in Polymer Science).
  • Eco-efficiency improvements in the propylene-to-epichlorohydrin process: This research provides an analysis of improving the eco-efficiency of the propylene-to-epichlorohydrin production process, which is crucial for industrial applications like windmill blade manufacture (Ł Madej, AA Kiss, 2023, Journal of Chemical Technology & Biotechnology).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.178 - 1.182
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

82.4 °F

Flash Point(C)

28 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service