Skip to Content
MilliporeSigma

8.03209

1,1-Dichloroethylene

(stabilised with hydroquinone monomethyl ether) for synthesis

Synonym(s):

1,1-Dichloroethylene, Vinylidene chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Cl2C=CH2
CAS Number:
Molecular Weight:
96.94
UNSPSC Code:
12352101
EC Index Number:
200-864-0
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

665 hPa ( 20 °C)

Quality Segment

assay

≥99.0% (GC)

form

liquid

autoignition temp.

520 °C

expl. lim.

8.4-16.5 % (v/v)

bp

31.2-31.7 °C/1013 hPa

mp

-122 °C

transition temp

flash point -25 °C

solubility

2.5 g/L

density

1.21 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

ClC(=C)Cl

InChI

1S/C2H2Cl2/c1-2(3)4/h1H2

InChI key

LGXVIGDEPROXKC-UHFFFAOYSA-N

Application

  • Thermodynamics and Kinetics for the Dechlorination of 1, 1-Dichloroethylene by Pd/Fe Nanoparticles: This study discusses the dechlorination mechanisms and the efficiency of Pd/Fe nanoparticles, offering insights into the thermodynamic and kinetic aspects of the process. (Ji et al., 2015).
  • Kinetics of the Aerobic Co-metabolism of 1,1-Dichloroethylene: Research on the aerobic degradation of 1,1-Dichloroethylene by a novel benzene-grown culture, providing insights into environmental bioremediation strategies. (Wang et al., 2014).
  • Discovery of an Inducible Toluene Monooxygenase That Co-oxidizes 1,4-Dioxane and 1,1-Dichloroethylene: This study explores the biodegradation capabilities of a specific strain of Azoarcus, shedding light on its potential for cleaning up sites contaminated with chlorinated solvents. (Deng et al., 2020).
  • Theoretical Investigations of Difluorobenzene Interacting with Ne and Infrared Analysis of 1, 1-Dichloroethylene from Synchrotron Data: Analyzes the infrared spectroscopy data of 1,1-Dichloroethylene, contributing to better molecular understanding through high-resolution spectroscopy. (Kamari, 2016).
  • Highly Selective Gas-Phase Synthesis of 1, 1-Dichloroethylene from 1, 1, 2-Trichloroethane Over Supported Amine Catalysts: Discusses a novel method for the synthesis of 1,1-Dichloroethylene, highlighting advancements in chemical manufacturing processes. (Tang et al., 2015).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Identity (IR): passes test
Density (d 5°C/ 4°C): 1.238 - 1.242
The material is stabilised with hydroquinone monomethyl ether.
At time of analytical testing a content of >100 ppm stabiliser was determined.


Still not finding the right product?

Explore all of our products under 1,1-Dichloroethylene


signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 1 - STOT RE 1 Inhalation - STOT RE 2 Oral

target_organs

Liver, Nose

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

-9.4 °F - closed cup

flash_point_c

-23.0 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library