Skip to Content
MilliporeSigma

8.03060

N,N-Dimethylaniline

for synthesis

Synonym(s):

N,N-Dimethylaniline, Dimethylaminobenzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5N(CH3)2
CAS Number:
Molecular Weight:
121.18
UNSPSC Code:
12352116
EC Index Number:
204-493-5
NACRES:
NA.22
MDL number:
Assay:
≥99% (GC)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.53 hPa ( 20 °C)

Quality Segment

assay

≥99% (GC)

form

liquid

autoignition temp.

370 °C (DIN 51794)

potency

1120 mg/kg LD50, oral (Rat), 1700 mg/kg LD50, skin (Rabbit)

expl. lim.

1.2-7 % (v/v)

pH

7.4 (20 °C, 1.2 g/L in H2O)

bp

192-195 °C/1013 hPa

mp

2.5 °C

transition temp

flash point 75 °C

solubility

1.2 g/L

density

0.96 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(C)(C)c1ccccc1

InChI

1S/C8H11N/c1-9(2)8-6-4-3-5-7-8/h3-7H,1-2H3

InChI key

JLTDJTHDQAWBAV-UHFFFAOYSA-N

Application

<ul>
<li><strong>Dual Fluorescence Bioimaging:</strong> N,N-Dimethylaniline derivatives to develop NIR-II emissive donor-acceptor-donor fluorophores. These compounds are used for dual fluorescence bioimaging and photothermal therapy applications, illustrating the critical role of N,N-Dimethylaniline in the development of new biomedical imaging agents (Sparks et al., 2024).</li>
<li><strong>Real-Time Monitoring of Biomolecules: </strong>Dimethylaniline is used in the construction of a fluorescent probe for in vivo real-time imaging and inhibition of A and beta;-fibril formation (Chen et al., 2024).</li>
<li><strong><strong>Photoinduced Electron Transfer Studies: </strong></strong>N,N-dimethylaniline is used in the quenching of the S1 state of two polar dyes, coumarin 152a and Nile red (Verma et al., 2024).</li>
</ul>

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.955 - 0.957
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under N,N-Dimethylaniline


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

167.0 °F - closed cup

flash_point_c

75 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library