Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.03052

Sigma-Aldrich

Dimethylammonium chloride

for synthesis

Synonym(s):

Dimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NH · HCl
CAS Number:
Molecular Weight:
81.54
MDL number:
UNSPSC Code:
12352107
EC Index Number:
208-046-5
NACRES:
NA.22

vapor pressure

<0.1 hPa ( 25 °C)

Quality Level

form

crystals

potency

1070 mg/kg LD50, oral (Rat)

pH

5 (20 °C, 100 g/L in H2O)

mp

160 °C

bulk density

500 kg/m3

storage temp.

2-30°C

InChI

1S/C2H7N.ClH/c1-3-2;/h3H,1-2H3;1H

InChI key

IQDGSYLLQPDQDV-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 98.0 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service