Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.02676

Sigma-Aldrich

Cyclododecanone

for synthesis

Synonym(s):

Cyclododecanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O
CAS Number:
Molecular Weight:
182.30
MDL number:
UNSPSC Code:
12352115
EC Index Number:
212-595-6

vapor pressure

<1 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

crystals

autoignition temp.

230 °C

potency

>2500 mg/kg LD50, oral (Rat)

bp

277 °C/1013 hPa

mp

59-61 °C

transition temp

flash point 118 °C

density

0.97 g/cm3 at 20 °C

InChI

1S/C12H22O/c13-12-10-8-6-4-2-1-3-5-7-9-11-12/h1-11H2

InChI key

SXVPOSFURRDKBO-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 59 °C
Melting range (upper value): ≤ 61 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service