Skip to Content
MilliporeSigma

8.02491

1-Cyanoguanidine

for synthesis

Synonym(s):

1-Cyanoguanidine, Dicyanodiamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
NCNHC(=NH)NH2
CAS Number:
Molecular Weight:
84.08
UNSPSC Code:
12352117
EC Index Number:
207-312-8
NACRES:
NA.22
MDL number:
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

potency

>5000 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rabbit)

mp

208-210 °C

density

1.4 g/cm3 at 20 °C

bulk density

400 kg/m3

storage temp.

2-30°C

SMILES string

NC(=NC#N)N

InChI

1S/C2H4N4/c3-1-6-2(4)5/h(H4,4,5,6)

InChI key

QGBSISYHAICWAH-UHFFFAOYSA-N

Analysis Note

Assay (ex N): ≥ 98.0 %
Melting range (lower value): ≥ 206 °C
Melting range (upper value): ≤ 210 °C
Identity (IR): passes test


pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
802491100004022536373775
802491999904022536373782
802491905004022536761244
802491010004022536373768