Skip to Content
MilliporeSigma

8.02256

2,2′-Biphenyldicarboxylic acid

for synthesis

Synonym(s):

2,2′-Biphenyldicarboxylic acid, Diphenic acid, Diphenyl-2,2-dicarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
2-(HOOC)C6H4C6H4-2-(COOH)
CAS Number:
Molecular Weight:
242.23
UNSPSC Code:
12352106
EC Index Number:
207-576-4
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (acidimetric)

autoignition temp.

390 °C

pH

4-5 (20 °C in H2O)

mp

225-228 °C

bulk density

470 kg/m3

storage temp.

2-30°C

SMILES string

OC(=O)c1c(cccc1)c2c(cccc2)C(=O)O

InChI

1S/C14H10O4/c15-13(16)11-7-3-1-5-9(11)10-6-2-4-8-12(10)14(17)18/h1-8H,(H,15,16)(H,17,18)

InChI key

GWZCCUDJHOGOSO-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Melting range (lower value): ≥ 225 °C
Melting range (upper value): ≤ 228 °C
Identity (IR): passes test


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library