Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C7H7NO
CAS Number:
Molecular Weight:
121.14
UNSPSC Code:
12352111
EC Index Number:
200-227-7
NACRES:
NA.22
MDL number:
Form:
crystals
vapor pressure
0.000056 hPa ( 20 °C)
Quality Segment
form
crystals
autoignition temp.
>500 °C
potency
1125 mg/kg LD50, oral (Rat)
pH
6.9 ( in H2O, saturated solution)
mp
124-127 °C
transition temp
flash point 180 °C
solubility
13 g/L
density
1.34 g/cm3 at 20 °C
bulk density
550 kg/m3
storage temp.
2-30°C
SMILES string
[N-H]C(=[O+H])c1ccccc1
InChI
1S/C7H7NO/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H2,8,9)
InChI key
KXDAEFPNCMNJSK-UHFFFAOYSA-N
Analysis Note
Assay (ex N): ≥ 99.0 %
Melting range (lower value): ≥ 124 °C
Melting range (upper value): ≤ 127 °C
Identity (IR): passes test
Melting range (lower value): ≥ 124 °C
Melting range (upper value): ≤ 127 °C
Identity (IR): passes test
Still not finding the right product?
Explore all of our products under Benzamide
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Muta. 2
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
356.0 °F - closed cup
flash_point_c
180 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 8021910250 | 04022536372037 |
| 8021910005 | 04022536372020 |
| 8021919025 | 04022536918990 |

