Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.01549

Sigma-Aldrich

1-Bromo-2-methylpropane

for synthesis

Synonym(s):

1-Bromo-2-methylpropane, iso-Butyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2Br
CAS Number:
Molecular Weight:
137.02
MDL number:
UNSPSC Code:
12352101
EC Index Number:
201-141-2
NACRES:
NA.22

vapor pressure

41 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

>2000 mg/kg LD50, oral (Rat)

bp

92 °C/1013 hPa

mp

-118 °C

transition temp

flash point 19 °C

solubility

slightly soluble 0.51 g/L

density

1.26 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H9Br/c1-4(2)3-5/h4H,3H2,1-2H3

InChI key

HLVFKOKELQSXIQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.262 - 1.264
Identity (IR): passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service