Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.00742

Sigma-Aldrich

1,2,3,6-Tetrahydrophthalic anhydride

for synthesis

Synonym(s):

1,2,3,6-Tetrahydrophthalic anhydride, 4-Cyclohexene-1,2-dicarboxylic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O3
CAS Number:
Molecular Weight:
152.15
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-605-4
NACRES:
NA.22

vapor pressure

0.00021 hPa ( 20 °C)

Quality Level

form

flakes

autoignition temp.

400 °C

potency

5410 mg/kg LD50, oral (Rat)

pH

2.1 (20 °C, 10 g/L in H2O)

mp

101.4 °C

transition temp

flash point 156 °C

solubility

29.4 g/L

density

1.36 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O1C(=O)C2C(CC=CC2)C1=O

InChI

1S/C8H8O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h1-2,5-6H,3-4H2

InChI key

KMOUUZVZFBCRAM-UHFFFAOYSA-N

Analysis Note

Assay (morpholine method): ≥ 97.0 %
Melting range (lower value): ≥ 99 °C
Melting range (upper value): ≤ 102 °C
Identity (IR): passes test

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

312.8 °F

Flash Point(C)

156 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service