Skip to Content
MilliporeSigma

8.00585

Butyl methacrylate

(stabilised) for synthesis

Synonym(s):

Butyl methacrylate, Butyl-2-methylpropenoate, Methacrylic acid butyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH2=C(CH3)COO(CH2)3CH3
CAS Number:
Molecular Weight:
142.20
UNSPSC Code:
12352108
EC Index Number:
202-615-1
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

2.7 hPa ( 20 °C)

Quality Segment

assay

≥99.0% (GC)

form

liquid

autoignition temp.

290 °C

potency

>2000 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rabbit)

expl. lim.

2.00-8.00 % (v/v)

bp

163 °C/1013 hPa

mp

-75 °C

transition temp

flash point 46 °C

density

0.89 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCCC)C(=O)C(=C)C

InChI

1S/C8H14O2/c1-4-5-6-10-8(9)7(2)3/h2,4-6H2,1,3H3

InChI key

SOGAXMICEFXMKE-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.894 - 0.896
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under Butyl methacrylate


pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

119.3 °F - closed cup

flash_point_c

48.5 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library