Skip to Content
MilliporeSigma

8.00508

Palmitic acid

for synthesis

Synonym(s):

Palmitic acid, Hexadecanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)14COOH
CAS Number:
Molecular Weight:
256.42
UNSPSC Code:
12352106
EC Index Number:
200-312-9
NACRES:
NA.22
MDL number:
Assay:
≥98% (GC)
Form:
solid
Grade:
synthesis grade
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Segment

assay

≥98% (GC)

form

solid

potency

>10000 mg/kg LD50, oral (Rat)

bp

271.4 °C/133 hPa

mp

61-63 °C

transition temp

flash point 206 °C

density

0.852 g/cm3 at 20 °C

bulk density

415 kg/m3

storage temp.

2-30°C

SMILES string

[O-]C(=O)CCCCCCCCCCCCCCC.[H+]

InChI

1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)

InChI key

IPCSVZSSVZVIGE-UHFFFAOYSA-N

Application

<ul>
<li><strong>Palmitic Acid in Biochemical Imaging:</strong> Research highlighted the use of palmitic acid derivatives in two-photon fluorescence imaging to investigate cellular mechanisms in abdominal aortic aneurysm, demonstrating its utility in advanced medical diagnostics and research (Liang et al., 2024).</li>
</ul>

Analysis Note

Assay (GC, area%): ≥ 98 %
Melting range
- lower value:
≥ 62 °C
- upper value: ≤ 64 °C
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under Palmitic acid


Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

235.4 °F

flash_point_c

113 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library