Skip to Content
MilliporeSigma

Skip To

8.00412

Chloroacetic acid

for synthesis

Synonym(s):

Chloroacetic acid, Monochloroacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C2H3ClO2
CAS Number:
Molecular Weight:
94.50
UNSPSC Code:
12352106
EC Index Number:
201-178-4
NACRES:
NA.22
MDL number:
Assay:
≥99% (acidimetric)
Form:
crystals
Grade:
synthesis grade
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Segment

vapor pressure

0.02 hPa ( 20 °C)

assay

≥99% (acidimetric)

form

crystals

autoignition temp.

460 °C

potency

90.4 mg/kg LD50, oral (Rat), 250 mg/kg LD50, skin (Rabbit)

expl. lim.

8.0 % (v/v)

pH

<1 (20 °C, 800 g/L in H2O)

mp

63 °C

transition temp

flash point 126 °C

solubility

>1000 g/L

density

1.64 g/cm3 at 20 °C

bulk density

800 kg/m3

storage temp.

2-30°C

SMILES string

ClCC(=O)O

InChI

1S/C2H3ClO2/c3-1-2(4)5/h1H2,(H,4,5)

InChI key

FOCAUTSVDIKZOP-UHFFFAOYSA-N

Application

  • Synthesis, characterization and study of the biological activity of some new compounds containing Triazine and 1, 3, 4-Thiadiazol units: Describes the synthesis of Carboxylic acid compound from Chloroacetic acid, highlighting its application in forming biologically active compounds (Abbas et al., 2023).
  • Extraction and Characterization of Cellulose from Broccoli Stems as a New Biopolymer Source for Producing Carboxymethyl Cellulose Films: Focuses on the carboxymethylation reaction involving Chloroacetic acid for producing carboxymethyl cellulose films, providing insights into biopolymer applications (Sayanjali et al., 2024).

Analysis Note

Assay (acidimetric): ≥ 99,0 %(m)
Melting range (lower value): ≥ 60 °C
Melting range (upper value): ≤ 65 °C
Identity (IR): conforms

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
40292305-2810C19627
grade

synthesis grade

grade

ACS reagent

grade

JIS special grade

grade

-

assay

≥99% (acidimetric)

assay

≥99.0%

assay

≥99.0%

assay

99%

Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

200

form

crystals

form

solid

form

crystalline

form

crystals

solubility

>1000 g/L

solubility

water: soluble 3170 g/L at 10 °C

solubility

water: soluble 3170 g/L at 10 °C

solubility

water: soluble 3,170 g/L at 10 °C

storage temp.

2-30°C

storage temp.

-

storage temp.

-

storage temp.

-


Still not finding the right product?

Explore all of our products under Chloroacetic acid


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

258.8 °F - closed cup

flash_point_c

126 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters