Skip to Content
MilliporeSigma

8.00238

Ethyl cinnamate

for synthesis

Synonym(s):

Ethyl cinnamate, Ethyl trans-3-phenylpropenoate, Cinnamic acid ethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5CH=CHCOOC2H5
CAS Number:
Molecular Weight:
176.21
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.22
Assay:
≥98.0% (GC)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (GC)

form

liquid

bp

271 °C/1013 hPa

mp

6-7 °C

transition temp

flash point 135 °C

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC)C(=O)\C=C\c1ccccc1

InChI

1S/C11H12O2/c1-2-13-11(12)9-8-10-6-4-3-5-7-10/h3-9H,2H2,1H3/b9-8+

InChI key

KBEBGUQPQBELIU-CMDGGOBGSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.048 - 1.050
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.


Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

275.0 °F

Flash Point (°C)

135 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library