Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.00140

Sigma-Aldrich

Benzilic acid

for synthesis

Synonym(s):

Benzilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2C(OH)COOH
CAS Number:
Molecular Weight:
228.24
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-993-2
NACRES:
NA.22

Quality Level

Assay

≥98.0% (acidimetric)

form

powder

autoignition temp.

530 °C

bp

180 °C/17.3 hPa

mp

150-152 °C

storage temp.

2-30°C

SMILES string

OC(c2ccccc2)(c1ccccc1)C(=O)O

InChI

1S/C14H12O3/c15-13(16)14(17,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,17H,(H,15,16)

InChI key

UKXSKSHDVLQNKG-UHFFFAOYSA-N

Application



  • Synthesis and characterization of various organic fluorescent and thermally stable polyesters with s-triazine frame and bulky side chain: This study highlights the use of benzilic acid as a bulky pendant group contributing to the fluorescence and thermal stability of polyesters (JV Modh, SA Pillai - CURRENT SCIENCE, 2023). Access the paper.


  • Simultaneous Determination of 13 α-Hydroxy Acids in Cosmetics by Ultra-High Performance Liquid Chromatography Triple Quadrupole Mass Spectrometry: Discusses the quantification of benzilic acid among other α-hydroxy acids in cosmetics, highlighting its importance in skincare products (高天阳 et al. - Physical Testing and Chemical Analysis Part B: Chemical Analysis, 2023). Access the paper.


  • Exploiting benzilic acid as a modular template: controlling photoreactivity and solid to liquid transition during photodimerization: This research uses benzilic acid as a core component to explore controlled photoreactivity and phase transitions in synthesized materials (MR Ahsan et al. - Chemical Communications, 2023). Access the paper.


  • Multicomponent Cyclizative 1, 2‐Rearrangement Enabled Enantioselective Construction of 2, 2‐Disubstituted Pyrrolinones: Reports a novel benzilic acid-type rearrangement in the synthesis of complex organic molecules, enhancing the enantioselectivity of the process (XZ Li et al. - Angewandte Chemie, 2024). Access the paper.




Analysis Note

Assay (acidimetric): ≥ 98.0 %
Melting range (lower value): ≥ 146 °C
Melting range (upper value): ≤ 150 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service