Skip to Content
MilliporeSigma

8.00086

Diethyl adipate

for synthesis

Synonym(s):

Diethyl adipate, Diethyl hexanedioate, Adipic acid diethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C2H5OOC(CH2)4COOC2H5
CAS Number:
Molecular Weight:
202.25
UNSPSC Code:
12352108
EC Index Number:
205-477-0
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99.0% (GC)

form

liquid

bp

245 °C/1013 hPa

mp

-19.8 °C

transition temp

flash point >100 °C

density

1.01 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC)C(=O)CCCCC(=O)OCC

InChI

1S/C10H18O4/c1-3-13-9(11)7-5-6-8-10(12)14-4-2/h3-8H2,1-2H3

InChI key

VIZORQUEIQEFRT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.007 - 1.009
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.


Still not finding the right product?

Explore all of our products under Diethyl adipate


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library