Skip to Content
MilliporeSigma

8.00028

Acetophenone

for synthesis

Synonym(s):

Acetophenone, Methyl phenyl ketone, 1-Phenylethanone, Phenylethanone, ACP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H8O
CAS Number:
Molecular Weight:
120.15
UNSPSC Code:
12352115
EC Index Number:
202-708-7
NACRES:
NA.22
MDL number:
eCl@ss:
39023804
Assay:
≥99.0% (GC)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.45 hPa ( 25 °C)

Quality Segment

assay

≥99.0% (GC)

form

liquid

autoignition temp.

570 °C

potency

815 mg/kg LD50, oral (Rat), 16000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.4-5.2 % (v/v)

transition temp

flash point 105 °C

density

1.03 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C(C)c1ccccc1

InChI

1S/C8H8O/c1-7(9)8-5-3-2-4-6-8/h2-6H,1H3

InChI key

KWOLFJPFCHCOCG-UHFFFAOYSA-N

Analysis Note

Colour (visual): colorless to yellowish
Appearance of substance (visual): clear liquid
Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.025 - 1.028
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.


Still not finding the right product?

Explore all of our products under Acetophenone


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

179.6 °F - closed cup

flash_point_c

82 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library