Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73026BC

Millipore

GDS Adhesive Strips

BioControl, For use with GDS

Synonym(s):

Sealing film

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121814

form

film

manufacturer/tradename

BioControl Systems 73026

storage condition

room temperature

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS Rotor-Gene

storage temp.

room temp

General description

GDS Adhesive Strips are the consumable accessories of the Assurance® GDS assays. Assurance® GDS is an automated nucleic acid amplification system for the detection of pathogenic organisms in foods, ingredients, and environmental samples. The sealing film strip is 19.0 x 97.3 mm, including two removable positioning tabs. The long dimension with tabs removed is 76.2 mm. These adhesive sheets are a reliable method of sealing wells for PCR applications. The high integrity sealing materials give the best protection against evaporation for high efficiency PCR.

Application

The Assurance® GDS Adhesive Strips are sealing film strips to provide effective and airtight sealing and help protect PCR samples during sample preparation.

Features and Benefits

  • Strong adhesive prevents evaporation during thermal cycling or sample storage
  • No sticky residue on the plate after the strip is peeled off
  • End tabs for easy seal application and removal
  • Clear for sample visualization and inspection
  • Temperature range: -20°C to 120°C

Components

Box of 200 film strips (50 sheets of four strips)

Compatibility

BioControl GDS

Testing Method

PCR, Immunomagnetic Separation (IMS)

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service