Skip to Content
MilliporeSigma
All Photos(1)

Documents

71401-M

Sigma-Aldrich

Rosetta 2(DE3)pLysS Singles Competent Cells - Novagen

Escherichia coli, rod shaped

Synonym(s):

BL21 derivatives

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106202
NACRES:
NA.81

product name

Rosetta 2(DE3)pLysS Singles Competent Cells - Novagen, Novagen′s Rosetta 2 (pLysS) host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli along with T7 lysozyme activity.

biological source

Escherichia coli

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

transformation efficiency: >2 × 106 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

Genotype: F-ompT hsdSB(rB- mB-) gal dcm (DE3) pLysSRARE2 (CamR)





This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
A common method for transformation of DNA plasmids into E. coli is the use of chemically competent cells. Although competent cells can be prepared in the laboratory, greater efficiency, reproducibility, and convenience are achieved using Novagen prepared competent cells. Novagen® Singles Competent Cells are designed for ultimate convenience and reliability in plasmid transformation. Chemically competent cells are prepared using an optimized method. Cells are provided in 50µl volumes, eliminating the need to aliquot, freeze/thaw, or waste partially used vials, saving time and money and ensuring reliable cell performance. To use, simply thaw, add DNA, incubate 5 minutes on ice, heat shock for 30 seconds, place on ice for 2 minutes, and add SOC medium.
T7 expression strains are lysogens of bacteriophageDE3, as indicated by the (DE3). These hosts carry achromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in appropriate T7 expression vectors, using IPTG as an inducer.
Rosetta 2 host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli. These strains supply tRNAs for 7 rare codones (AGA, AGG, AUA, CUA, GGA, CCC, and CGG) on a compatible chloramphenicol-resistant plasmid. The tRNA genes are driven by their native promoters.

DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.

pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
Novagen′s Rosetta 2 (pLysS) host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli along with T7 lysozyme activity.

Features and Benefits

• Guaranteed transformation efficiencies:

>1.5 × 10⁸ cfu/µg (NovaBlue andNovaBlueT1R)

>2 × 10⁶ cfu/µg [BL21(DE3), BL21(DE3)pLysS,Origami(DE3), Origami 2(DE3),Rosetta(DE3), Rosetta 2(DE3)pLysS]

• Provided as frozen single-use aliquots in 11-reaction or 22-reaction kits

• Includes Test Plasmid and SOC Medium

• Selection of popular host strains: NovaBlue forhigh efficiency cloning, and λDE3 lysogens forprotein expression with pET vectors

• Prepared using an optimized chemical method

Components

•11 rxn or 22 rxnRosetta 2(DE3)pLysS Competent Cells

•2 × 2 ml or 4 × 2 mlSOC Medium

•10 µlTest Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service