Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

71238-M

Sigma-Aldrich

pIEx-2 DNA - Novagen

Novagen′s pIEx vectors are designed for cloning and high-level expression of proteins in transiently transfected Spodoptera derived insect cells.

Synonym(s):

InsectDirect System- pIEx-2 DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106611
NACRES:
NA.51

Quality Level

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze

storage temp.

−20°C

General description

The pIEx vectors are designed for cloning and high-level expression of proteins in transiently transfected Spodoptera-derived insect cells. Transcription is driven by the AcNPV-derived hr5 enhancer and immediate early promoter, IE1. pIEx-2 contains N-terminal GST•Tag, His•Tag, and S•Tag coding sequences, and a C-terminal HSV•Tag coding sequence. N-terminal vector encoded sequences can be completely removed by cloning into the PshA I site and cleaving the fusion protein with enterokinase.
pIEx-2 is also available as an Ek/LIC vector. The Ek/LIC vector is prepared for rapid directional cloning of PCR-amplified DNA. Using specifically designed primers for amplification, inserts can be efficiently cloned without the need for restriction enzyme digestion or ligation.

For sequencing or PCR of recombinants in pIEx-2, refer to the table below to choose the appropriate primers.

Warning

Toxicity: Standard Handling (A)

Legal Information

INSECTDIRECT is a trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service