Skip to Content
MilliporeSigma
All Photos(1)

Documents

71135

Sigma-Aldrich

Rosetta-gami B Competent Cells - Novagen

Escherichia coli, rod shaped

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106202
NACRES:
NA.81

product name

Rosetta-gami B Competent Cells - Novagen, Rosetta-gami B strains combine the key features of BL21, Origami, and Rosetta to enhance both the expression of eukaryotic proteins and the formation of target protein disulfide bonds.

biological source

Escherichia coli

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

shipped in

dry ice

storage temp.

−70°C

General description

Genotype: FompT hsdSB (rB mB) gal dcm lacY1 ahpC gor522::Tn10 trxB pRARE (CamR, KanR, TetR)





This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
Rosetta-gami B strains combine the key features of BL21 (and its Tuner derivative), Origami, and Rosetta to enhance both the expression of eukaryotic proteins and the formation of target protein disulfide bonds in the bacterial cytoplasm. These strains are compatible with ampicillin- or spectinomycin-resistant vectors.
Rosetta-gami B strains combine the key features of BL21, Origami, and Rosetta to enhance both the expression of eukaryotic proteins and the formation of target protein disulfide bonds.

Components

0.4 ml1 mlComponent

•2 × 0.2 ml5 × 0.2 mlRosetta-gami B Competent Cells

•2 × 2 ml4 × 2 mlSOC Medium

•10 µl10 µlTest Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service