Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.51
UNSPSC Code:
41106616
Form:
liquid
Storage temp.:
2-8°C
form
liquid
Quality Level
manufacturer/tradename
Novagen®
storage condition
OK to freeze, avoid repeated freeze/thaw cycles
shipped in
wet ice
storage temp.
2-8°C
Legal Information
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
T7SELECT is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 70040-3 | 04055977273922 |