Skip to Content
MilliporeSigma

69872-M

pET-29b(+) DNA - Novagen

Novagen′s pET-29a-c(+) vectors carry an N-terminal S•Tag/thrombin configuration plus an optional C-terminal His•Tag® sequence.

Synonym(s):

pET System, pET-29a-c(+) vectors, pET-29b(+) DNA Vector

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.51
UNSPSC Code:
41106610
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

manufacturer/tradename

Novagen®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

shipped in

dry ice, wet ice

storage temp.

−70°C

General description

The pET-29a-c(+) vectors carry an N-terminal S•Tag/thrombin configuration plus an optional C-terminal HIS TAG® sequence. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map (TB076). The f1 origin is oriented so that infection with helper phage will produce virions containing single-stranded DNA that corresponds to the coding strand.. The pET Vectors are supplied as purified plasmid DNA (10 µg). Each order of pET DNA also includes an Induction Control strain (supplied as a glycerol stock). Please contact technical service if you need additional information.

Application

Novagen′s pET-29a-c(+) vectors carry an N-terminal S•Tag/thrombin configuration plus an optional C-terminal HIS TAG® sequence.

Legal Information

HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany
HSV-TAG is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

This product is sold for internal research use only. Any commercial use of this product, its components, and/or any derivatives thereof (including but not limited to proteins produced using the product or its components) (together and hereinafter the ′EMD Product′) requires signature of a written commercial use agreement with EMD Millipore Corporation or its successor-in-interest. Commercial use shall include but not be limited to: (1) use of the EMD Product to manufacture products for sale to third parties; (2) use of the EMD Product to provide services, information, or data to third parties in exchange for consideration; (3) use of the EMD Product for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the EMD Product, whether or not such EMD Product is resold for research use. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of the EMD Product. Please direct any questions on these use restrictions to: licensing@milliporesigma.com.

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges MilliporeSigma to request certain information about you and the establishment where the GMOs are being handled. Clickhere for Enduser Declaration (EUD) Form
Toxicity: Standard Handling


Still not finding the right product?

Explore all of our products under pET-29b(+) DNA - Novagen


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library