Skip to Content
MilliporeSigma
All Photos(1)

Documents

69240-M

Sigma-Aldrich

S. cerevisiae Genomic DNA

High molecular weight, high purity DNA for any application

Synonym(s):

Saccharomyces cerevisiae DNA, Yeast Genomic DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352208
NACRES:
NA.51

Quality Level

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

2-8°C

General description

Genomic DNAs are highly purified preparations of high molecular weight genomic DNA from a variety of organisms. The isolation protocol minimizes shearing and removes contaminants that inhibit enzymatic procedures. Novagen genomic DNA is qualified for genomic analysis (including PCR) and library construction. At least 90% of the DNA is greater than 100 kbp in size, as analyzed by CHEF gel electrophoresis (>50 kbp for S. cerevisiae). No inhibition of restriction enzyme activity is observed, and the DNA is free of contaminating RNA and protein.

The DNA is supplied in 10 mM Tris-HCl, pH 8.0, 1 mM EDTA (TE). Pipet with a wide-borepipet tip to minimize shearing.

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service