Skip to Content
Merck
All Photos(1)

Key Documents

69182

Millipore

DNase I, RNase free

For applications in which maintenance of RNA integrity is critical

Synonym(s):

RNase-free DNase I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.85

Pricing and availability is not currently available.

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

technique(s)

DNA extraction: suitable

suitability

suitable for nucleic acid purification

application(s)

diagnostic assay manufacturing

shipped in

wet ice

storage temp.

−20°C

General description

RNase-free DNase I digests either single- or double-stranded DNA, producing a mixture of mono- and oligonucleotides. Purified to be free of RNase, this preparation is qualified for applications in which maintenance of the integrity of RNA is critical. The enzyme selectively degrades DNA in the presence of RNA and can be used to remove DNA template following in vitro transcription reactions. This enzyme is also useful in other applications such as DNase footprinting and nick translation.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit will degrade 1 μg DNA in 10 minutes at 37°C. The reaction mixture (50 µl) contains 80 mM HEPES pH 7.5, 10 mM NaCl, 5 mM MgCl₂, 10 mM DTT, 1 µg plasmid DNA, and enzyme.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service