Skip to Content
MilliporeSigma

69026

T7•Tag® Antibody Agarose

Synonym(s):

T7 Tag Antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.56
UNSPSC Code:
41106500
Clone:
-
Species reactivity:
-
Application:
Citations:
8
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

manufacturer/tradename

Novagen®

storage condition

do not freeze

shipped in

wet ice

storage temp.

2-8°C

General description

The T7•Tag Antibody Agarose is designed for rapid immunoaffinity purification of target proteins that carry the T7•Tag sequence (i.e., the amino terminal 11 aa of the T7 gene 10 protein). Purification is based on binding target proteins to T7•Tag monoclonal antibody that is covalently coupled to cross-linked agarose beads, washing away unbound proteins, and eluting at pH 2.2. Capacity will vary somewhat between different target proteins, but the beads are standardized to bind a minimum of 300 µg T7•Tag β-galactosidase per milliliter of settled resin. The beads can be used in either batch or column methods and can be recycled a minimum of five times without loss of binding activity.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
T7-Tag is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library