Skip to Content
MilliporeSigma

677181

Vincristine Sulfate

≥98% (TLC), Microtuble disruptor, solid

Synonym(s):

Vincristine Sulfate, Apocynaceae sp.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C46H56N4O10 · xH2SO4
CAS Number:
Molecular Weight:
824.96 (free base basis)
UNSPSC Code:
12352210
NACRES:
NA.77
MDL number:
Assay:
≥98% (TLC)
Form:
solid
Storage condition:
OK to freeze
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Vincristine Sulfate, Apocynaceae sp., Indole alkaloid that binds to tubulin, inhibiting formation of microtubules.

Quality Segment

description

Merck USA index - 14, 9986

assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 20 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)([O-])[O-].[N+H]21C[C@H](C[C@@](C2)(O)CC)C[C@@](c8[nH]c9c(c8CC1)cccc9)(c3cc4c(cc3OC)N([C@@H]5[C@]64[C@H]7[N+H](CC6)CC=C[C@]7([C@H]([C@]5(O)C(=O)OC)OC(=O)C)CC)C=O)C(=O)OC

InChI

1S/C46H56N4O10.H2O4S/c1-7-42(55)22-28-23-45(40(53)58-5,36-30(14-18-48(24-28)25-42)29-12-9-10-13-33(29)47-36)32-20-31-34(21-35(32)57-4)50(26-51)38-44(31)16-19-49-17-11-15-43(8-2,37(44)49)39(60-27(3)52)46(38,56)41(54)59-6;1-5(2,3)4/h9-13,15,20-21,26,28,37-39,47,55-56H,7-8,14,16-19,22-25H2,1-6H3;(H2,1,2,3,4)/t28-,37+,38-,39-,42+,43-,44-,45+,46+;/m1./s1

InChI key

AQTQHPDCURKLKT-JKDPCDLQSA-N

General description

Indole alkaloid that binds to tubulin, inhibiting formation of microtubules. Depolymerizes microtubules. Induces increase in micronuclei and delays cell cycle progression. Anti-neoplastic agent. Induces apoptosis in human lymphoma cells by disrupting microtubules.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no

Other Notes

Chan, M.W., et al. 1998. Cancer Biochem. Biophys. 16, 347.
Hanauske, A.R., et al. 1994. Eur. J. Cancer30A, 1688.
Takano, Y., et al. 1993. Pathol. Res. Pract. 189, 187.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Carcinogenic / Teratogenic (D)


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Muta. 2 - Repr. 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library