Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

676505

Sigma-Aldrich

VEGFR2 Kinase Inhibitor VII, SKLB1002

The VEGFR2 Kinase Inhibitor VII, SKLB1002 controls the biological activity of VEGFR2 Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

VEGFR2 Kinase Inhibitor VII, SKLB1002, 6,7-Dimethoxy-4-(5-methyl-1,3,4-thiadiazol-2-ylthio)quinazoline, Vascular Endothelial Growth Factor Receptor 2 Kinase Inhibitor VII, SKLB1002, VEGFR Tyrosine Kinase Inhibitor XXXV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12N4O2S2
Molecular Weight:
320.39
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 2.5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable quinazoline that acts as a potent, ATP-binding site-targeting, VEGFR2-selective inhibitor (IC50 = 32 nM), while exhibiting much reduced potency against c-kit, Ret, FMS, PDGFRα, and Aurora A (IC50 = 0.62, 2.5, 2.9, 3.1, and 3.9 µM, respectively) and little or no activity against 11 other kinases (IC50 >10 µM). An effective antiangiogenesis agent (86%, 92%, and 98% inhibition, respectively, of HUVEC migration, invasion, and tube formation at 10 µM; 80% inhibition of zebrafish embryos intersegmental vessel growth at 2.5 µM) that significantly suppresses SW620- and HepG2-derived tumor growth in mice (by 72% and 63%, respectively, in 18 days; 100 mg/kg/day; i.p.) in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Zhang, S., et al. 2011. Clin. Cancer Res.17, 4439.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service